ENA390552686 Enamine

3-phenylpyrrolidin-2-one

  • MDL number MFCD01687225
  • Cas Number : 6836-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82...84
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127536

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-127536
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart