ENA390552689 Enamine

methyl 2-methanesulfonylpropanoate

  • MDL number MFCD16045024
  • Cas Number : 73017-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127540

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127540
Availability details for
Quantity
  • Availability
  • Add to Cart