ENA390552692 Enamine

2-amino-N-(3-bromophenyl)acetamide

  • MDL number MFCD09724303
  • Cas Number : 938338-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79...81
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127545

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127545
Availability details for
Quantity
  • Availability
  • Add to Cart