ENA390552693 Enamine

2-bromo-6-(1,3-dioxolan-2-yl)pyridine

  • MDL number MFCD06209635
  • Cas Number : 34199-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127546

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127546
Availability details for
Quantity
  • Availability
  • Add to Cart