ENA390552695 Enamine

2-(2-phenoxyethoxy)propanoic acid

  • MDL number MFCD09939916
  • Cas Number : 1016817-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127550

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127550
Availability details for
Quantity
  • Availability
  • Add to Cart