ENA390552697 Enamine

2-chloro-3-(propan-2-yloxy)pyrazine

  • MDL number MFCD14702666
  • Cas Number : 1209457-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127555

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127555
Availability details for
Quantity
  • Availability
  • Add to Cart