ENA390552698 Enamine

4-(oxolan-3-yl)piperidine

  • MDL number MFCD11040367
  • Cas Number : 1211528-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126183
Availability details for
Quantity
  • Availability
  • Add to Cart