ENA390552707 Enamine

2-(oxan-2-yl)acetonitrile

  • MDL number MFCD08234895
  • Cas Number : 75394-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127597

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127597
Availability details for
Quantity
  • Availability
  • Add to Cart