ENA390552708 Enamine

3-(4-chloropyrimidin-2-yl)phenol

  • MDL number MFCD12107760
  • Cas Number : 1155596-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126236

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126236
Availability details for
Quantity
  • Availability
  • Add to Cart