ENA390552716 Enamine

(2-cyanophenyl)methyl acetate

  • MDL number MFCD20265196
  • Cas Number : 38866-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-127608

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127608
Availability details for
Quantity
  • Availability
  • Add to Cart