ENA390552721 Enamine

2-methyl-3-[(pyrrolidin-1-yl)(thiophen-2-yl)methyl]-1H-indole

  • MDL number MFCD04092285
  • Cas Number : 618407-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-127619

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127619
Availability details for
Quantity
  • Availability
  • Add to Cart