ENA390552724 Enamine

1-(3,4-dichlorophenyl)pyrrolidin-3-amine

  • MDL number MFCD12164694
  • Cas Number : 1183558-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127637

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127637
Availability details for
Quantity
  • Availability
  • Add to Cart