ENA390552728 Enamine

ethyl 4,5-difluoro-1H-indole-2-carboxylate

  • MDL number MFCD11007943
  • Cas Number : 194870-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127645
Availability details for
Quantity
  • Availability
  • Add to Cart