ENA390552729 Enamine

4-sulfanylnaphthalen-1-ol

  • MDL number MFCD19301479
  • Cas Number : 45993-54-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127646

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127646
Availability details for
Quantity
  • Availability
  • Add to Cart