ENA390552735 Enamine

2-(tert-butylsulfanyl)pyridine-4-carbonitrile

  • MDL number MFCD11136355
  • Cas Number : 501074-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127662

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127662
Availability details for
Quantity
  • Availability
  • Add to Cart