ENA390552736 Enamine

3-[(4-fluorophenyl)amino]-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD00174653
  • Cas Number : 324570-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127664

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127664
Availability details for
Quantity
  • Availability
  • Add to Cart