ENA390552742 Enamine

methyl 2-{[(benzyloxy)carbonyl]amino}-3-[(4-methylbenzenesulfonyl)oxy]propanoate

  • MDL number MFCD24369222
  • Cas Number : 21142-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127672

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127672
Availability details for
Quantity
  • Availability
  • Add to Cart