ENA390552743 Enamine

(4-propylphenyl)methanol

  • MDL number MFCD06201146
  • Cas Number : 82657-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127673
Availability details for
Quantity
  • Availability
  • Add to Cart