ENA390552746 Enamine

2-fluoro-3-phenylpropanoic acid

  • MDL number MFCD01722162
  • Cas Number : 457-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71-74
Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127676

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127676
Availability details for
Quantity
  • Availability
  • Add to Cart