ENA390552750 Enamine

5-bromo-2-(2-methoxyethoxy)aniline

  • MDL number MFCD08687012
  • Cas Number : 946664-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127682
Availability details for
Quantity
  • Availability
  • Add to Cart