ENA390552751 Enamine

1-(propan-2-yl)-1H-imidazole-4,5-dicarbonitrile

  • MDL number MFCD18808570
  • Cas Number : 133123-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127683

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127683
Availability details for
Quantity
  • Availability
  • Add to Cart