ENA390552752 Enamine

4-(4-chlorophenyl)-3-methyl-4-oxobutanoic acid

  • MDL number MFCD00156903
  • Cas Number : 52240-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73...75
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126368

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126368
Availability details for
Quantity
  • Availability
  • Add to Cart