ENA390552753 Enamine

ethyl 3-(tert-butylamino)propanoate

  • MDL number MFCD01475854
  • Cas Number : 1462-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-127690

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127690
Availability details for
Quantity
  • Availability
  • Add to Cart