ENA390552754 Enamine

2-(4-bromo-2-methylphenyl)acetonitrile

  • MDL number MFCD14582920
  • Cas Number : 215800-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127692
Availability details for
Quantity
  • Availability
  • Add to Cart