ENA390552760 Enamine

2,3-dibromopyrazine

  • MDL number MFCD02684897
  • Cas Number : 95538-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127698

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127698
Availability details for
Quantity
  • Availability
  • Add to Cart