ENA390552766 Enamine

6-methyl-2,3,4,9-tetrahydro-1H-carbazol-3-one

  • MDL number MFCD21188571
  • Cas Number : 1432682-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155...157
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127707

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127707
Availability details for
Quantity
  • Availability
  • Add to Cart