ENA390552767 Enamine

N-(5-bromo-1,3-thiazol-2-yl)-4-methoxybenzene-1-sulfonamide

  • MDL number MFCD12720599
  • Cas Number : 1183841-34-0
  • Molecular Weight : 349.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-126375

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126375
Availability details for
Quantity
  • Availability
  • Add to Cart