ENA390552769 Enamine

[(1-benzyl-1H-pyrazol-4-yl)methyl](propan-2-yl)amine

  • MDL number MFCD12403336
  • Cas Number : 1153115-77-5
  • Molecular Weight : 229.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-127718

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127718
Availability details for
Quantity
  • Availability
  • Add to Cart