ENA390552771 Enamine

benzyl[(4-propoxyphenyl)methyl]amine hydrochloride

  • MDL number MFCD15163625
  • Cas Number : 1158513-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219...221
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127727

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127727
Availability details for
Quantity
  • Availability
  • Add to Cart