ENA390552788 Enamine

1-(5-bromo-2-hydroxyphenyl)-2,2-dimethylpropan-1-one

  • MDL number MFCD16742948
  • Cas Number : 1247482-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127782

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127782
Availability details for
Quantity
  • Availability
  • Add to Cart