ENA390552792 Enamine

tert-butyl 4-(pyridazin-3-yl)piperazine-1-carboxylate

  • MDL number MFCD19689624
  • Cas Number : 492431-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-127787

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127787
Availability details for
Quantity
  • Availability
  • Add to Cart