ENA390552795 Enamine

5,6-dichloro-3H-imidazo[4,5-b]pyridine

  • MDL number MFCD20681407
  • Cas Number : 189102-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127793

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127793
Availability details for
Quantity
  • Availability
  • Add to Cart