ENA390552796 Enamine

1-(3-bromopropyl)-3-methylbenzene

  • MDL number MFCD09744407
  • Cas Number : 103324-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127795

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127795
Availability details for
Quantity
  • Availability
  • Add to Cart