ENA390552801 Enamine

1-[4-methyl-2-(propan-2-yl)-1,3-thiazol-5-yl]ethan-1-ol

  • MDL number MFCD16112734
  • Cas Number : 1250760-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Country of Origin Ukraine

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127804
Availability details for
Quantity
  • Availability
  • Add to Cart