ENA390552807 Enamine

4-(4-formyl-1-methyl-1H-pyrazol-3-yl)benzonitrile

  • MDL number MFCD11180177
  • Cas Number : 1152508-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156...158
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127819

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127819
Availability details for
Quantity
  • Availability
  • Add to Cart