ENA390552811 Enamine

N-(naphthalen-1-ylmethyl)aniline

  • MDL number MFCD01570883
  • Cas Number : 7182-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54...56
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126479

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126479
Availability details for
Quantity
  • Availability
  • Add to Cart