ENA390552814 Enamine

2-isocyanato-1-benzothiophene

  • MDL number MFCD14660418
  • Cas Number : 124808-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127846
Availability details for
Quantity
  • Availability
  • Add to Cart