ENA390552818 Enamine

1-(2-chlorophenyl)-1-methoxypropan-2-amine

  • MDL number MFCD16062871
  • Cas Number : 1092403-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127856

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127856
Availability details for
Quantity
  • Availability
  • Add to Cart