ENA390552821 Enamine

1-(1-benzylpyrrolidin-3-yl)ethan-1-amine

  • MDL number MFCD22057269
  • Cas Number : 107259-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127860

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127860
Availability details for
Quantity
  • Availability
  • Add to Cart