ENA390552824 Enamine

2-amino-3-[(2-methoxy-2-oxoethyl)sulfanyl]propanoic acid

  • MDL number MFCD09944188
  • Cas Number : 51124-86-8
  • Molecular Weight : 193.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126490

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126490
Availability details for
Quantity
  • Availability
  • Add to Cart