ENA390552831 Enamine

3-(benzyloxy)-2-fluoropyridine

  • MDL number MFCD23837834
  • Cas Number : 1432680-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34...36
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127898

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127898
Availability details for
Quantity
  • Availability
  • Add to Cart