ENA390552834 Enamine

2-(morpholin-4-yl)-3-nitrobenzoic acid

  • MDL number MFCD06809611
  • Cas Number : 890091-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196...198
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126532

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126532
Availability details for
Quantity
  • Availability
  • Add to Cart