ENA390552840 Enamine

2-(2-acetamidoethoxy)acetic acid

  • MDL number MFCD21933536
  • Cas Number : 765249-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126555

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126555
Availability details for
Quantity
  • Availability
  • Add to Cart