ENA390552843 Enamine

2-(3-phenylpropyl)-2,3-dihydro-1H-inden-1-ol

  • MDL number MFCD16517683
  • Cas Number : 1285167-20-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127922

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127922
Availability details for
Quantity
  • Availability
  • Add to Cart