ENA390552852 Enamine

methyl 1-cyano-2-methylcyclobutane-1-carboxylate

  • MDL number MFCD21773094
  • Cas Number : 1432679-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127950

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127950
Availability details for
Quantity
  • Availability
  • Add to Cart