ENA390552854 Enamine

4-bromo-2-chloro-6-methylbenzoic acid

  • MDL number MFCD19686179
  • Cas Number : 877149-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127953

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127953
Availability details for
Quantity
  • Availability
  • Add to Cart