ENA390552858 Enamine

4-(4-bromophenyl)-1-phenylbutan-1-one

  • MDL number MFCD16461419
  • Cas Number : 1225857-78-2
  • Molecular Weight : 303.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(4-bromophenyl)-1-phenyl-1-butanone
Inchi Code 1S/C16H15BrO/c17-15-11-9-13(10-12-15)5-4-8-16(18)14-6-2-1-3-7-14/h1-3,6-7,9-12H,4-5,8H2
Inchi Key JVHPRBJJKZQBPR-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-127959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127959
Availability details for
Quantity
  • Availability
  • Add to Cart