ENA390552861 Enamine

2-(trifluoroacetamido)benzoic acid

  • MDL number MFCD00175073
  • Cas Number : 19165-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167...169
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127963

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127963
Availability details for
Quantity
  • Availability
  • Add to Cart