ENA390552862 Enamine

3-methanesulfonylprop-2-enoic acid

  • MDL number MFCD20692347
  • Cas Number : 63068-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127964
Availability details for
Quantity
  • Availability
  • Add to Cart