ENA390552863 Enamine

2-(2-aminoethoxy)-N-methylacetamide hydrochloride

  • MDL number MFCD24369118
  • Cas Number : 1432678-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-126575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126575
Availability details for
Quantity
  • Availability
  • Add to Cart